Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
709-1317
RL709-1317EA 292.54 USD
RL709-1317
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
F(ab')2 Anti-Human Fc antibody is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.

Recommended Dilutions: ELISA: 1:130,000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR