Anti-IgL Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
709-1111
RL709-1111EA 292.54 USD
RL709-1111
Anti-IgL Goat Polyclonal Antibody
Antibodies
Suitable for highly specific immunological methods requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:6000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Light Chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR