Anti-IgG Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Rockland Immunochemical
706-408-002
RL706-408-002EA 329.1 USD
RL706-408-002
Anti-IgG Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
Recommended Dilutions:
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR