Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
702-403-002
RL702-403-002EA 265.37 USD
RL702-403-002
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Antibody anti-Cat has been assayed against 1.0 µg of Cat IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:5000 - 1:100000; Immunochemistry: 1:500 - 1:5000;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR