F(ab')2 Anti-Biotin Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemical
700-105-098
RL700-105-098EA 690.6 USD
RL700-105-098
F(ab')2 Anti-Biotin Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
This product has been assayed against 1.0 µg of Biotin conjuagted IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:1000
Do not freeze!

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR