Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemical
619-105-002
RL619-105-002EA 292.54 USD
RL619-105-002
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended Dilutions: ELISA: 1:2000 - 1:25000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Armenian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR