Anti-IgG Rabbit Polyclonal Antibody (DyLight® 649)

Supplier: Rockland Immunochemical
613-443-002-0.5
RL6134430020.5EA 438.48 USD
RL6134430020.5
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 649)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 649
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR