Anti-IgG Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
612-9302
RL612-9302EA 258.06 USD
RL612-9302
Anti-IgG Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Rat IgG antibody, Peroxidase Conjugate has been assayed against 1.0 µg of Rat IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:10000 - 1:100000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR