Anti-DNM1 Sheep Polyclonal Antibody

Supplier: Rockland Immunochemical
612-601-D33
RL612-601-D33EA 656.12 USD
RL612-601-D33
Anti-DNM1 Sheep Polyclonal Antibody
Antibodies
Anti-Dynamin pS778 Antibody is suitable for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 95 kDa in size corresponding to dynamin Protein phosphorylated at Ser778.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000

Type: Primary
Antigen: DNM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR