Anti-SLC32A1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
612-401-E53
RL612-401-E53EA 656.12 USD
RL612-401-E53
Anti-SLC32A1 Rabbit Polyclonal Antibody
Antibodies
Anti-VGAT antibody is suitable for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 53 kDa in size corresponding to vesicular GABA transporter Protein.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000

Type: Primary
Antigen: SLC32A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR