Anti-SLC6A13 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
612-401-D57
RL612-401-D57EA 656.12 USD
RL612-401-D57
Anti-SLC6A13 Rabbit Polyclonal Antibody
Antibodies
Anti-GAT2 (Rabbit) Antibody is suitable for use in IHC. This antibody is not recommended for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. GAT2 antibody is specific for the approximately 82 kDa in size.

Recommended Dilutions: Immunochemistry: 1:100-200

Type: Primary
Antigen: SLC6A13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR