Anti-PPP1R1B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
612-401-D24
RL612-401-D24EA 656.12 USD
RL612-401-D24
Anti-PPP1R1B Rabbit Polyclonal Antibody
Antibodies
Anti-DARPP-32 (Rabbit) Antibody is suitable for use in ELISA, Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 32 kDa in size corresponding to DARPP 32 Protein in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000; Immunochemistry: 1:250

Type: Primary
Antigen: PPP1R1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR