Anti-IgA Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
612-1106
RL612-1106EA 206.87 USD
RL612-1106
Anti-IgA Goat Polyclonal Antibody
Antibodies
Anti-Rat IgA antibody is suitable for Elisa, Western Blots and immunochemistry. Antibody should be optimized by the end user for specific conditions of reactivity.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:3000;

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR