Anti-IgG Sheep Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemical
611-6502
RL611-6502EA 228.81 USD
RL611-6502
Anti-IgG Sheep Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Anti-Rabbit IgG antibody is suitable for ELISA, Western Blot and immunochemistry applications. Specific conditions for reactivity should be optimized by the researcher.

Recommended Dilutions: ELISA: 1:15000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Sheep
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR