Anti-IgG Sheep Polyclonal Antibody (DyLight® 680)

Supplier: Rockland Immunochemical
611-644-122
RL611-644-122EA 147.31 USD
RL611-644-122
Anti-IgG Sheep Polyclonal Antibody (DyLight® 680)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope: Heavy and Light chain
Host: Sheep
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR