Anti-IgG Sheep Polyclonal Antibody

Supplier: Rockland Immunochemical
611-6102
RL611-6102EA 143.13 USD
RL611-6102
Anti-IgG Sheep Polyclonal Antibody
Antibodies
Anti-RABBIT IgG (H&L) (Sheep) Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:40000 - 1:200,000; Western Blot: 1:4000 - 1:20000; Immunochemistry: 1:2000 - 1:10000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Sheep
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR