Anti-IgG Guinea pig Polyclonal Antibody

Supplier: Rockland Immunochemical
611-201-122
RL611-201-122EA 228 USD
RL611-201-122
Anti-IgG Guinea pig Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Guinea Pig
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR