Anti-IgG Rabbit Polyclonal Antibody (DyLight® 405)

Supplier: Rockland Immunochemical
610-446-002-0.5
RL6104460020.5EA 463.88 USD
RL6104460020.5
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 405)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR