Anti-IgM µ Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
610-4107-0500
RL6104107-0500EA 83.58 USD
RL6104107-0500
Anti-IgM µ Rabbit Polyclonal Antibody
Antibodies
Anti-Mouse IgM mu heavy chain antibody is suitable for highly specific immunological methods requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:500 - 1:2500; Other Dilution: User Optimized

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR