Anti-IgG Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
610-406-C46
RL610406C46EA 268.51 USD
RL610406C46
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Antibodies
Anti-Mouse IgG subclass pan reactive Secondary Antibody is suitable for highly specific immunological methods requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR