Anti-IgG Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
610-1619-0500
RL6101619-0500EA 112.84 USD
RL6101619-0500
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR