Anti-IgG Sheep Polyclonal Antibody (DyLight® 549)

Supplier: Rockland Immunochemical
610-642-002
RL610-642-002EA 143.13 USD
RL610-642-002
Anti-IgG Sheep Polyclonal Antibody (DyLight® 549)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope: Heavy and Light chain
Host: Sheep
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR