Anti-IgG2a Rabbit Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemical
610-4941
RL610-4941EA 268.51 USD
RL610-4941
Anti-IgG2a Rabbit Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Rabbit Anti-IgG2a Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR