Anti-IgG2b gamma Rabbit Polyclonal Antibody (DyLight® 405)

Supplier: Rockland Immunochemical
610-446-042
RL610-446-042EA 143.13 USD
RL610-446-042
Anti-IgG2b gamma Rabbit Polyclonal Antibody (DyLight® 405)
Antibodies
Type: Secondary
Antigen: IgG2b gamma
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR