Anti-IgG gamma RabbitPolyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
610-4312
RL610-4312EA 311.34 USD
RL610-4312
Anti-IgG gamma RabbitPolyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Rabbit Anti-IgG (gamma chain) Reacts with Mouse

Recommended Dilutions: ELISA: 1:21000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: gamma chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR