Anti-IgG2b gamma Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
610-4142
RL610-4142EA 228.81 USD
RL610-4142
Anti-IgG2b gamma Rabbit Polyclonal Antibody
Antibodies
Secondary Rabbit Anti-IgG2b Reacts with Mouse

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG2b gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR