Anti-IgG Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
610-1602
RL610-1602EA 177.61 USD
RL610-1602
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Mouse IgG (H&L) biotin conjugated antibody is suitable for highly specific immunological methods requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:350000; Western Blot: 1:2000 - 1:20000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR