Anti-IgG Goat Polyclonal Antibody (Cy5.5®)

Supplier: Rockland Immunochemical
610-113-121
RL610-113-121EA 338 USD
RL610-113-121
Anti-IgG Goat Polyclonal Antibody (Cy5.5®)
Antibodies
Secondary Goat Anti-IgG (H&L) Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR