Anti-IgM µ Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
610-1107
RL610-1107EA 238.21 USD
RL610-1107
Anti-IgM µ Goat Polyclonal Antibody
Antibodies
Antibody Anti-Mouse IgM (mu chain) is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR