Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
610-106-430
RL610-106-430EA 283.13 USD
RL610-106-430
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)
Antibodies
Immunoblotting, ELISA, immunochemistry, immunomicroscopy as well as other antibody based assays using streptavidin or avidin conjugates requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:300,000; Western Blot: 1:3000 - 1:30,000; Immunochemistry: 1:1500 - 1:15000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR