Anti-IgG3 gamma Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemical
610-102-043
RL610-102-043EA 268.9 USD
RL610-102-043
Anti-IgG3 gamma Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Anti-Mouse IgG3 secondary antibody is suitable for Flow Cytometry, immunochemistry and Immunofluorescence Microscopy, ELISA, western blotting as well as other anti IgG2b antibody based assays.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:200 - 1:500; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG3 gamma
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR