Anti-IgG Chicken Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemical
609-9502
RL609-9502EA 289.4 USD
RL609-9502
Anti-IgG Chicken Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
This product has been assayed against 1.0 µg of Human IgG in a standard capture ELISA using pNPP (p-nitrophenyl phosphate) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR