Anti-IgG Donkey Polyclonal Antibody (Cy2®)

Supplier: Rockland Immunochemical
609-711-123
RL609-711-123EA 322.84 USD
RL609-711-123
Anti-IgG Donkey Polyclonal Antibody (Cy2®)
Antibodies
Recommended Dilutions: FLISA: 1:10000 - 1:50000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR