Anti-IgG Rabbit Polyclonal Antibody (DyLight® 549)

Supplier: Rockland Immunochemical
609-442-002
RL609-442-002EA 155.67 USD
RL609-442-002
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 549)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR