Anti-IgG gamma Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemical
609-4212
RL609-4212EA 240.3 USD
RL609-4212
Anti-IgG gamma Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Secondary Rabbit Anti-IgG (gamma chain) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: gamma chain
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR