Anti-IgG gamma Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemical
609-1912
RL609-1912EA 240.3 USD
RL609-1912
Anti-IgG gamma Goat Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Goat Anti-IgG (gamma chain) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: gamma chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR