Anti-IgL Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemical
609-1911
RL609-1911EA 289.4 USD
RL609-1911
Anti-IgL Goat Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Goat Anti-λ (lambda chain) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Light Chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR