Anti-IgA alpha Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
609-1606
RL609-1606EA 192.24 USD
RL609-1606
Anti-IgA alpha Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-HUMAN IgA (alpha chain) (GOAT) Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoelectron microscopy and immunochemistry as well as other antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:60,000 - 1:600,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: alpha chain
Host: Goat
Isotype: IgA
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR