Anti-IgG Goat Polyclonal Antibody (DyLight® 680)

Supplier: Rockland Immunochemical
609-144-002
RL609-144-002EA 155.67 USD
RL609-144-002
Anti-IgG Goat Polyclonal Antibody (DyLight® 680)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR