Anti-Ig kappa Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
609-1310
RL609-1310EA 314.48 USD
RL609-1310
Anti-Ig kappa Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-human kappa chain Antibody has been assayed against 1.0 µg of human kappa light chain in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:77000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: Igkappa
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR