Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
609-1302
RL609-1302EA 214.18 USD
RL609-1302
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Human IgG (H&L) peroxidase conjugated antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:180,000; Western Blot: 1:5000 - 1:25000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR