Anti-IgL Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
609-1111
RL609-1111EA 217.31 USD
RL609-1111
Anti-IgL Goat Polyclonal Antibody
Antibodies
Suitable for immunoblotting (western or dot blot), ELISA, immunoelectron microscopy and immunochemistry as well as other antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Light Chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR