Anti-IgA/IgG/IgM Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
609-101-130
RL609-101-130EA 206.87 USD
RL609-101-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Antibodies
Secondary Goat Anti-IgG IgA IgM Reacts with Human

Recommended Dilutions: ELISA: 1:70,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR