Anti-IgG Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
606-1103
RL606-1103EA 174.48 USD
RL606-1103
Anti-IgG Goat Polyclonal Antibody
Antibodies
Anti-Guinea Pig IgG F(c) antibody has been assayed against 1.0 µg of Sheep IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR