Anti-IgG Donkey Polyclonal Antibody (DyLight® 549)

Supplier: Rockland Immunochemical
605-742-125
RL605742125EA 147.31 USD
RL605742125
Anti-IgG Donkey Polyclonal Antibody (DyLight® 549)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR