Anti-IgG Donkey Polyclonal Antibody (DyLight® 488)

Supplier: Rockland Immunochemical
605-741-002-0.5
RL6057410020.5EA 478.51 USD
RL6057410020.5
Anti-IgG Donkey Polyclonal Antibody (DyLight® 488)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR