Anti-IgG Rabbit Polyclonal Antibody (DyLight® 680)

Supplier: Rockland Immunochemical
605-444-002-0.5
RL6054440020.5EA 463.88 USD
RL6054440020.5
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 680)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR