Anti-IgG Donkey Polyclonal Antibody

Supplier: Rockland Immunochemical
605-701-125
RL605-701-125EA 174.48 USD
RL605-701-125
Anti-IgG Donkey Polyclonal Antibody
Antibodies
Secondary Donkey Anti-IgG (H&L) Reacts with Goat

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR