Anti-IgG Mouse Polyclonal Antibody

Supplier: Rockland Immunochemical
605-301-002
RL605-301-002EA 177.61 USD
RL605-301-002
Anti-IgG Mouse Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Mouse
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR