Anti-IgG Goat Polyclonal Antibody (DyLight® 649)

Supplier: Rockland Immunochemical
603-143-126
RL603143126EA 147.31 USD
RL603143126
Anti-IgG Goat Polyclonal Antibody (DyLight® 649)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 649
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR